Learn More
Cytokeratin 7-Specific Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16857-1-AP-20UL
This item is not returnable.
View return policy
Description
The Cytokeratin 7-specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Cytokeratin 7-specific. This antibody recognizes human, mouse antigen. The Cytokeratin 7-specific antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Cytokeratin 7-Specific | |
Polyclonal | |
Unconjugated | |
20 μL | |
3855 | |
IgG |
Western Blot, Immunohistochemistry | |
180 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |