Learn More
Cyclohexyldiphenylphosphine, 98%, Thermo Scientific Chemicals
42.00 € - 1403.00 €
Chemical Identifiers
CAS | 6372-42-5 |
---|---|
Molecular Formula | C18H21P |
Molecular Weight (g/mol) | 268.34 |
MDL Number | MFCD00046360 |
InChI Key | ZXKWUYWWVSKKQZ-UHFFFAOYSA-N |
Synonym | cyclohexyldiphenylphosphine, phosphine, cyclohexyldiphenyl, cyclohexyl diphenyl phosphine, diphenylcyclohexylphosphine, diphenylphosphinocyclohexane, cyclohexyl-diphenyl-phosphane, pubchem6451, acmc-1ba4g, cyclohexyl diphenyl phosphane |
PubChem CID | 80756 |
SMILES | C1CCC(CC1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15451467
|
Thermo Scientific Alfa Aesar
036708.03 |
1 g | |||||||
11334237
|
Thermo Scientific Alfa Aesar
036708.06 |
5 g |
112.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11324237
|
Thermo Scientific Alfa Aesar
036708.14 |
25 g | |||||||
11344237
|
Thermo Scientific Alfa Aesar
036708.22 |
100 g |
1403.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclohexyldiphenylphosphine is used as ligand in coupling reaction. It is also used as a medical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6372-42-5 | |
268.34 | |
ZXKWUYWWVSKKQZ-UHFFFAOYSA-N | |
80756 |
C18H21P | |
MFCD00046360 | |
cyclohexyldiphenylphosphine, phosphine, cyclohexyldiphenyl, cyclohexyl diphenyl phosphine, diphenylcyclohexylphosphine, diphenylphosphinocyclohexane, cyclohexyl-diphenyl-phosphane, pubchem6451, acmc-1ba4g, cyclohexyl diphenyl phosphane | |
C1CCC(CC1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
6372-42-5 | |
>110°C (230°F) | |
MFCD00046360 | |
cyclohexyldiphenylphosphine, phosphine, cyclohexyldiphenyl, cyclohexyl diphenyl phosphine, diphenylcyclohexylphosphine, diphenylphosphinocyclohexane, cyclohexyl-diphenyl-phosphane, pubchem6451, acmc-1ba4g, cyclohexyl diphenyl phosphane | |
ZXKWUYWWVSKKQZ-UHFFFAOYSA-N | |
cyclohexyl(diphenyl)phosphane | |
80756 | |
98% | |
Cyclohexyldiphenylphosphine |
58°C to 62°C | |
C18H21P | |
Air Sensitive | |
Insoluble in water. | |
C1CCC(CC1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
268.34 | |
268.34 | |
Solid |
Safety and Handling
P260-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P314-P330-P333+P313-P362-P501c
H302-H315-H317-H319-H373
missing translation for 'einecsNumber' : 228-904-2
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only