missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyldiphenylphosphine, 98%, Thermo Scientific Chemicals

Product Code. 11324237
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cyclohexyldiphenylphosphine is used as ligand in coupling reaction. It is also used as a medical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6372-42-5
Molecular Formula C18H21P
Molecular Weight (g/mol) 268.34
MDL Number MFCD00046360
InChI Key ZXKWUYWWVSKKQZ-UHFFFAOYSA-N
Synonym cyclohexyldiphenylphosphine, phosphine, cyclohexyldiphenyl, cyclohexyl diphenyl phosphine, diphenylcyclohexylphosphine, diphenylphosphinocyclohexane, cyclohexyl-diphenyl-phosphane, pubchem6451, acmc-1ba4g, cyclohexyl diphenyl phosphane
PubChem CID 80756
SMILES C1CCC(CC1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 58°C to 62°C
Flash Point >110°C (230°F)
Quantity 25 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
IUPAC Name cyclohexyl(diphenyl)phosphane
Formula Weight 268.34
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Cyclohexyldiphenylphosphine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'