Learn More
Cyclohexanone cyanohydrin, 98%, Thermo Scientific Chemicals
27.70 € - 416.00 €
Chemical Identifiers
CAS | 931-97-5 |
---|---|
Molecular Formula | C7H11NO |
Molecular Weight (g/mol) | 125.171 |
MDL Number | MFCD00003818 |
InChI Key | ZDBRPNZOTCHLSP-UHFFFAOYSA-N |
Synonym | cyclohexanone cyanohydrin, 1-hydroxycyclohexanecarbonitrile, 1-hydroxy-1-cyclohexanecarbonitrile, cyclohexanone, cyanohydrin, 1-cyano-1-hydroxycyclohexane, cyclohexanecarbonitrile, 1-hydroxy, unii-on43942n5p, ccris 4608, 1-hydroxy-cyclohexanecarbonitrile, cyclohexanoncyanhydrin |
PubChem CID | 13610 |
IUPAC Name | 1-hydroxycyclohexane-1-carbonitrile |
SMILES | C1CCC(CC1)(C#N)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11302649
|
Thermo Scientific Alfa Aesar
H26699.03 |
1 g |
27.70 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11312649
|
Thermo Scientific Alfa Aesar
H26699.09 |
10 g | N/A | ||||||
11322649
|
Thermo Scientific Alfa Aesar
H26699.18 |
50 g |
416.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
931-97-5 | |
125.171 | |
ZDBRPNZOTCHLSP-UHFFFAOYSA-N | |
13610 | |
C1CCC(CC1)(C#N)O |
C7H11NO | |
MFCD00003818 | |
cyclohexanone cyanohydrin, 1-hydroxycyclohexanecarbonitrile, 1-hydroxy-1-cyclohexanecarbonitrile, cyclohexanone, cyanohydrin, 1-cyano-1-hydroxycyclohexane, cyclohexanecarbonitrile, 1-hydroxy, unii-on43942n5p, ccris 4608, 1-hydroxy-cyclohexanecarbonitrile, cyclohexanoncyanhydrin | |
1-hydroxycyclohexane-1-carbonitrile |
Specifications
931-97-5 | |
132°C (19 mmHg) | |
C7H11NO | |
UN3439 | |
ZDBRPNZOTCHLSP-UHFFFAOYSA-N | |
1-hydroxycyclohexane-1-carbonitrile | |
13610 | |
98% |
34°C to 36°C | |
118°C (244°F) | |
MFCD00003818 | |
cyclohexanone cyanohydrin, 1-hydroxycyclohexanecarbonitrile, 1-hydroxy-1-cyclohexanecarbonitrile, cyclohexanone, cyanohydrin, 1-cyano-1-hydroxycyclohexane, cyclohexanecarbonitrile, 1-hydroxy, unii-on43942n5p, ccris 4608, 1-hydroxy-cyclohexanecarbonitrile, cyclohexanoncyanhydrin | |
C1CCC(CC1)(C#N)O | |
125.171 | |
125.17 | |
Cyclohexanone cyanohydrin |
Safety and Handling
GHS H Statement
H300
Fatal if swallowed.
GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
missing translation for 'rtecsNumber' : GU7710000
missing translation for 'tsca' : TSCA
Recommended Storage : 2° to 8°C
RUO – Research Use Only