missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone cyanohydrin, 98%, Thermo Scientific Chemicals

Product Code. 11322649
Click to view available options
Quantity:
1 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 931-97-5
Molecular Formula C7H11NO
Molecular Weight (g/mol) 125.171
MDL Number MFCD00003818
InChI Key ZDBRPNZOTCHLSP-UHFFFAOYSA-N
Synonym cyclohexanone cyanohydrin, 1-hydroxycyclohexanecarbonitrile, 1-hydroxy-1-cyclohexanecarbonitrile, cyclohexanone, cyanohydrin, 1-cyano-1-hydroxycyclohexane, cyclohexanecarbonitrile, 1-hydroxy, unii-on43942n5p, ccris 4608, 1-hydroxy-cyclohexanecarbonitrile, cyclohexanoncyanhydrin
PubChem CID 13610
IUPAC Name 1-hydroxycyclohexane-1-carbonitrile
SMILES C1CCC(CC1)(C#N)O

Specifications

Melting Point 34°C to 36°C
Density 1.031
Boiling Point 132°C (19 mmHg)
Flash Point 118°C (244°F)
Refractive Index 1.469
Quantity 50 g
UN Number UN3439
Formula Weight 125.17
Percent Purity 98%
Chemical Name or Material Cyclohexanone cyanohydrin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.