missing translation for 'onlineSavingsMsg'
Learn More

Cyclododecanone, 99%, Thermo Scientific™

26.00 € - 74.13 €

Chemical Identifiers

CAS 830-13-7
Molecular Formula C12H22O
Molecular Weight (g/mol) 182.307
MDL Number MFCD00003722
InChI Key SXVPOSFURRDKBO-UHFFFAOYSA-N
Synonym cyclododecan-1-one, dsstox_cid_7322, dsstox_rid_78407, dsstox_gsid_27322, cyclododecanon, cdon, cyclododecanone, acmc-209pr1, ksc448s4j
PubChem CID 13246
IUPAC Name cyclododecanone
SMILES C1CCCCCC(=O)CCCCC1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11322497
View Documents
Thermo Scientific Alfa Aesar
L11563.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11332497
View Documents
Thermo Scientific Alfa Aesar
L11563.36
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

830-13-7
182.307
SXVPOSFURRDKBO-UHFFFAOYSA-N
13246
C1CCCCCC(=O)CCCCC1
C12H22O
MFCD00003722
cyclododecan-1-one, dsstox_cid_7322, dsstox_rid_78407, dsstox_gsid_27322, cyclododecanon, cdon, cyclododecanone, acmc-209pr1, ksc448s4j
cyclododecanone

Specifications

830-13-7
277°C
Camphor-like
MFCD00003722
508621
SXVPOSFURRDKBO-UHFFFAOYSA-N
cyclododecanone
13246
99%
59°C to 62°C
137°C (278°F)
C12H22O
UN3077
cyclododecan-1-one, dsstox_cid_7322, dsstox_rid_78407, dsstox_gsid_27322, cyclododecanon, cdon, cyclododecanone, acmc-209pr1, ksc448s4j
C1CCCCCC(=O)CCCCC1
182.307
182.31
Cyclododecanone
Videos
Safety and Handling

Safety and Handling

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications