missing translation for 'onlineSavingsMsg'
Learn More

Cyclododecanone, 99%, Thermo Scientific™

Product Code. 11322497
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 830-13-7
Molecular Formula C12H22O
Molecular Weight (g/mol) 182.307
MDL Number MFCD00003722
InChI Key SXVPOSFURRDKBO-UHFFFAOYSA-N
Synonym cyclododecan-1-one, dsstox_cid_7322, dsstox_rid_78407, dsstox_gsid_27322, cyclododecanon, cdon, cyclododecanone, acmc-209pr1, ksc448s4j
PubChem CID 13246
IUPAC Name cyclododecanone
SMILES C1CCCCCC(=O)CCCCC1

Specifications

Melting Point 59°C to 62°C
Boiling Point 277°C
Flash Point 137°C (278°F)
Odor Camphor-like
Quantity 100 g
UN Number UN3077
Beilstein 508621
Formula Weight 182.31
Percent Purity 99%
Chemical Name or Material Cyclododecanone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'