Learn More
Copper(I) bromide-dimethyl sulfide complex, Thermo Scientific Chemicals
31.50 € - 386.00 €
Chemical Identifiers
CAS | 54678-23-8 |
---|---|
Molecular Formula | C2H6BrCuS |
Molecular Weight (g/mol) | 205.58 |
MDL Number | MFCD00043295 |
InChI Key | PMHQVHHXPFUNSP-UHFFFAOYSA-M |
Synonym | copper i bromide-dimethyl sulfide, bromo thiobis methane copper, copper i bromide dimethyl sulfide complex, bromocopper; methylsulfanylmethane, cubr.me2s, ksc493e2t, copper i bromide dimethylsulfide, copper i bromide-dimethylsulfide, bromo dimethylsulfide copper i |
PubChem CID | 2724234 |
SMILES | [Cu]Br.CSC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11333537
|
Thermo Scientific Alfa Aesar
019368.04 |
2 g |
31.50 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11343537
|
Thermo Scientific Alfa Aesar
019368.09 |
10 g |
93.30 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11323537
|
Thermo Scientific Alfa Aesar
019368.18 |
50 g |
386.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Copper(I) bromide-dimethyl sulfide complex is used as catalyst for adding Grignard reagents to fullerenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
54678-23-8 | |
205.58 | |
PMHQVHHXPFUNSP-UHFFFAOYSA-M | |
2724234 |
C2H6BrCuS | |
MFCD00043295 | |
copper i bromide-dimethyl sulfide, bromo thiobis methane copper, copper i bromide dimethyl sulfide complex, bromocopper; methylsulfanylmethane, cubr.me2s, ksc493e2t, copper i bromide dimethylsulfide, copper i bromide-dimethylsulfide, bromo dimethylsulfide copper i | |
[Cu]Br.CSC |
Specifications
124°C to 129°C (decomposition) | |
Brown or Green | |
C2H6BrCuS | |
UN3335 | |
Soluble in water. Reacts with water and decomposes. | |
[Cu]Br.CSC | |
205.58 | |
205.58 | |
Copper(I) bromide-dimethyl sulfide complex |
54678-23-8 | |
Crystalline | |
MFCD00043295 | |
copper i bromide-dimethyl sulfide, bromo thiobis methane copper, copper i bromide dimethyl sulfide complex, bromocopper; methylsulfanylmethane, cubr.me2s, ksc493e2t, copper i bromide dimethylsulfide, copper i bromide-dimethylsulfide, bromo dimethylsulfide copper i | |
PMHQVHHXPFUNSP-UHFFFAOYSA-M | |
bromocopper;methylsulfanylmethane | |
2724234 | |
Air and moisture sensitive |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.
missing translation for 'einecsNumber' : 259-292-5
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only