missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) bromide-dimethyl sulfide complex, Thermo Scientific Chemicals

Product Code. 11323537
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Copper(I) bromide-dimethyl sulfide complex is used as catalyst for adding Grignard reagents to fullerenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54678-23-8
Molecular Formula C2H6BrCuS
Molecular Weight (g/mol) 205.58
MDL Number MFCD00043295
InChI Key PMHQVHHXPFUNSP-UHFFFAOYSA-M
Synonym copper i bromide-dimethyl sulfide, bromo thiobis methane copper, copper i bromide dimethyl sulfide complex, bromocopper; methylsulfanylmethane, cubr.me2s, ksc493e2t, copper i bromide dimethylsulfide, copper i bromide-dimethylsulfide, bromo dimethylsulfide copper i
PubChem CID 2724234
SMILES [Cu]Br.CSC

Specifications

Melting Point 124°C to 129°C (decomposition)
Color Brown or Green
Physical Form Crystalline
Quantity 50 g
UN Number UN3335
Solubility Information Soluble in water. Reacts with water and decomposes.
IUPAC Name (methylsulfanyl)methane; bromocopper
Formula Weight 205.58
Sensitivity Air and moisture sensitive
Chemical Name or Material Copper(I) bromide-dimethyl sulfide complex

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'