missing translation for 'onlineSavingsMsg'
Learn More

COIL Mouse anti-Human, Unconjugated, Clone: 2G12B8, Proteintech

Mouse Monoclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen COIL
Clone 2G12B8
Concentration 500 μg/mL
Applications Western Blot, Immunofluorescence, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16809745
View Documents
Proteintech
67218-1-IG-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16899735
View Documents
Proteintech
67218-1-IG-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The COIL antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human COIL. This antibody recognizes human antigen. The COIL antibody has been validated for the following applications: IF, WB, ELISA analysis.

Specifications

Specifications

COIL
500 μg/mL
Monoclonal
Mouse
8161
Primary
2G12B8
Western Blot, Immunofluorescence, ELISA
Unconjugated
Human
IgG1
Videos
SDS
Documents

Documents

Product Certifications