Learn More
COIL Mouse anti-Human, Unconjugated, Clone: 2G12B8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67218-1-IG-150UL
This item is not returnable.
View return policy
Description
The COIL antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human COIL. This antibody recognizes human antigen. The COIL antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
COIL | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunofluorescence, ELISA | |
2G12B8 | |
Unconjugated | |
150 μL | |
8161 | |
IgG1 |