Learn More
Hirschmann™ Class B Graduated Polypropylene Cylinders with Raised Blue Graduations
Excellent for precise liquid measurement and fluid transfer in the laboratory.
46.75 € - 113.00 €
Specifications
Material Polypropylene
Certifications/Compliance DIN 12681, ISO 6706
Description Graduated cylinder, PP
Class B
Disposable Reusable
View More Specs
Products 6
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
13128029
View Documents Promotion Details
Hirschmann™
7500160
46.75 €
Pack of 12
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
13138029
View Documents Promotion Details
Hirschmann™
7500170
25 mL
52.90 €
Pack of 12
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
13148029
View Documents Promotion Details
Hirschmann™
7500175
60.30 €
Pack of 12
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
13168029
View Documents Promotion Details
Hirschmann™
7500186
57.85 €
Pack of 6
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
13188029
View Documents Promotion Details
Hirschmann™
7500193
1000 mL
99.65 €
Pack of 6
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
13198029
View Documents Promotion Details
Hirschmann™
7500194
113.00 €
Pack of 3
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12681/ISO 6706
  • PP
  • Raised scale
  • Main point ring graduation
  • Hexagonal base
Specifications
Specifications
Polypropylene
Graduated cylinder, PP
Reusable
Precise Liquid Measurement and Fluid Transfer in the Laboratory
Hexagonal
DIN 12681, ISO 6706
B
Round
Blue
Graduated Measuring Cylinder