Learn More
Hirschmann™ Class B Graduated Polypropylene Cylinders with Raised Blue Graduations
Excellent for precise liquid measurement and fluid transfer in the laboratory.
Brand:  Hirschmann™ 7500186
 View more versions of this product
Product Code. 13168029
  • 57.85 € / Pack of 6
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12681/ISO 6706
  • PP
  • Raised scale
  • Main point ring graduation
  • Hexagonal base
Specifications
Specifications
B
Round
Blue
315 mm
Reusable
Precise Liquid Measurement and Fluid Transfer in the Laboratory
Hexagonal
Graduated Measuring Cylinder