missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIB4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | CIB4 |
---|---|
Concentration | 0.29 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16858694
|
Proteintech
18840-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16848694
|
Proteintech
18840-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CIB4 (calciumand integrin-binding familymember 4) is a 185 amino acid protein thatcontainsthree EF-hand domains. CIB4 is closelyrelated to CIB (CIB has one less EF-hand domain),which is known to bind to Integrin alpha II beta in platelets and isinvolved in signal transduction. The gene encoding CIB4 maps to human chromosome 2,which houses over 1,400 genes and comprises nearly 8% of the human genome. Harlequin icthyosis, a rare andmorbid skin deformity, is associated with mutations in the ABCA12 gene,while the lipid metabolic disorders itosterolemia is associated with defectsin the ABCG5 and ABCG8 genes. Additionally, an extremely rare recessive genetic disorder, Alstrom syndrome, is caused by mutationsin the ALMS1 gene,which maps to chromosome 2.Specifications
CIB4 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
A0PJX0, Q9D9N5 | |
130106, 73259 | |
Peptide | |
Primary | |
-20°C | |
CIB4 |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CIB4, KIP4 | |
Cib4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title