missing translation for 'onlineSavingsMsg'
Learn More

CIB4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 18840-1-AP-20UL

 View more versions of this product

Product Code. 16858694

  • 145.00 € / 20µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

CIB4 (calciumand integrin-binding familymember 4) is a 185 amino acid protein thatcontainsthree EF-hand domains. CIB4 is closelyrelated to CIB (CIB has one less EF-hand domain),which is known to bind to Integrin alpha II beta in platelets and isinvolved in signal transduction. The gene encoding CIB4 maps to human chromosome 2,which houses over 1,400 genes and comprises nearly 8% of the human genome. Harlequin icthyosis, a rare andmorbid skin deformity, is associated with mutations in the ABCA12 gene,while the lipid metabolic disorders itosterolemia is associated with defectsin the ABCG5 and ABCG8 genes. Additionally, an extremely rare recessive genetic disorder, Alstrom syndrome, is caused by mutationsin the ALMS1 gene,which maps to chromosome 2.
Specifications

Specifications

CIB4
Polyclonal
Unconjugated
CIB4
CIB4, KIP4
Rabbit
Antigen Affinity Chromatography
RUO
130106, 73259
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A0PJX0, Q9D9N5
Cib4
Peptide
20 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.