Learn More
Chromium arsenide, 99% (metals basis), Thermo Scientific™
106.00 € - 405.00 €
Chemical Identifiers
CAS | 12254-85-2 |
---|---|
Molecular Formula | AsCr |
Molecular Weight (g/mol) | 126.92 |
MDL Number | MFCD00198045 |
InChI Key | OAMWSOUCVTVANH-UHFFFAOYSA-N |
Synonym | chromiumylidynearsane, chromium arsenide |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
12254-85-2 | |
126.92 | |
OAMWSOUCVTVANH-UHFFFAOYSA-N |
AsCr | |
MFCD00198045 | |
chromiumylidynearsane, chromium arsenide |
Specifications
12254-85-2 | |
AsCr | |
UN1557 | |
OAMWSOUCVTVANH-UHFFFAOYSA-N | |
178.98 | |
(metals basis) |
Powder | |
MFCD00198045 | |
chromiumylidynearsane, chromium arsenide | |
126.92 | |
99% | |
Chromium arsenide |
Safety and Handling
GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
missing translation for 'tsca' : TSCA