Learn More
Chlorotriisopropylsilane, 97+%, Thermo Scientific Chemicals
83.20 € - 1158.00 €
Chemical Identifiers
CAS | 13154-24-0 |
---|---|
Molecular Formula | C9H21ClSi |
Molecular Weight (g/mol) | 192.80 |
MDL Number | MFCD00009656 |
InChI Key | KQIADDMXRMTWHZ-UHFFFAOYSA-N |
Synonym | triisopropylsilyl chloride, chlorotriisopropylsilane, triisopropylchlorosilane, chlorotris propan-2-yl silane, chloro triisopropyl silane, silane, chlorotris 1-methylethyl, tipscl, triisopropylsilylchloride, triisopropyl chlorosilane, triisopropylchlorosilane, redistilled |
PubChem CID | 139400 |
IUPAC Name | chloro-tri(propan-2-yl)silane |
SMILES | CC(C)[Si](Cl)(C(C)C)C(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11483617
|
Thermo Scientific Alfa Aesar
A17376.09 |
10 g |
83.20 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11493617
|
Thermo Scientific Alfa Aesar
A17376.18 |
50 g |
264.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11403627
|
Thermo Scientific Alfa Aesar
A17376.30 |
250 g |
1158.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chlorotriisopropylsilane is used as a silylating agent in neucleotide synthesis. It is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals and in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChlorotriisopropylsilane is used as a silylating agent in neucleotide synthesis. It is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals and in organic synthesis.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents, strong bases, alcohols and amines.
Chemical Identifiers
13154-24-0 | |
192.80 | |
KQIADDMXRMTWHZ-UHFFFAOYSA-N | |
139400 | |
CC(C)[Si](Cl)(C(C)C)C(C)C |
C9H21ClSi | |
MFCD00009656 | |
triisopropylsilyl chloride, chlorotriisopropylsilane, triisopropylchlorosilane, chlorotris propan-2-yl silane, chloro triisopropyl silane, silane, chlorotris 1-methylethyl, tipscl, triisopropylsilylchloride, triisopropyl chlorosilane, triisopropylchlorosilane, redistilled | |
chloro-tri(propan-2-yl)silane |
Specifications
200°C | |
C9H21ClSi | |
UN2987 | |
62°C (144°F) | |
Miscible with water. | |
CC(C)[Si](Cl)(C(C)C)C(C)C | |
192.80 | |
192.81 | |
Acrid | |
Moisture sensitive | |
Chlorotriisopropylsilane |
13154-24-0 | |
MFCD00009656 | |
1737446 | |
triisopropylsilyl chloride, chlorotriisopropylsilane, triisopropylchlorosilane, chlorotris propan-2-yl silane, chloro triisopropyl silane, silane, chlorotris 1-methylethyl, tipscl, triisopropylsilylchloride, triisopropyl chlorosilane, triisopropylchlorosilane, redistilled | |
KQIADDMXRMTWHZ-UHFFFAOYSA-N | |
chloro-tri(propan-2-yl)silane | |
139400 | |
≥97% | |
1.4535 | |
0.908 |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.