missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorotriisopropylsilane, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Chlorotriisopropylsilane is used as a silylating agent in neucleotide synthesis. It is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals and in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chlorotriisopropylsilane is used as a silylating agent in neucleotide synthesis. It is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals and in organic synthesis.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents, strong bases, alcohols and amines.
Chlorotriisopropylsilane is used as a silylating agent in neucleotide synthesis. It is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals and in organic synthesis.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents, strong bases, alcohols and amines.
Specifications
Specifications
Boiling Point | 200°C |
Quantity | 250 g |
UN Number | UN2987 |
Beilstein | 1737446 |
Flash Point | 62°C (144°F) |
Solubility Information | Miscible with water. |
Formula Weight | 192.81 |
Percent Purity | ≥97% |
Odor | Acrid |
Refractive Index | 1.4535 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'