missing translation for 'onlineSavingsMsg'
Learn More

Chloranilic acid, 98+%, Thermo Scientific Chemicals

39.40 € - 424.11 €

Chemical Identifiers

CAS 87-88-7
Molecular Formula C6H2Cl2O4
Molecular Weight (g/mol) 208.978
MDL Number MFCD00001596
InChI Key IPPWILKGXFOXHO-UHFFFAOYSA-N
Synonym chloranilic acid, 2,5-dichloro-3,6-dihydroxy-p-benzoquinone, p-chloranilic acid, 2,5-cyclohexadiene-1,4-dione, 2,5-dichloro-3,6-dihydroxy, unii-yj8l3bb7y4, 2,5-dichloro-3,6-dihydroxybenzoquinone, 2,5-dihydroxy-3,6-dichlorobenzoquinone, yj8l3bb7y4, p-quinone, 2,5-dichloro-3,6-dihydroxy, p-benzoquinone, 2,5-dichloro-3,6-dihydroxy
PubChem CID 66604
IUPAC Name 2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione
SMILES C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11448733
View Documents
Thermo Scientific Alfa Aesar
A10493.14
25 g
39.40 €
25g
Please to purchase this item. Need a web account? Register with us today!
11458733
View Documents
Thermo Scientific Alfa Aesar
A10493.22
100 g
107.54 €
100g
Please to purchase this item. Need a web account? Register with us today!
11468733
View Documents
Thermo Scientific Alfa Aesar
A10493.36
500 g
424.11 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Chloranilic acid is used in spectrophotometry. It reacts with metal ions to form stable complexes. It is also used as a reactant in the preparation of dimethylbipyridyl complexes and (nonylbenzimidazolylmethyl)benzene. Further, it is used in charge-transfer reactions with metoprolol tartrate. It is also employed in the synthesis of osmium metalla-rectangles with anticancer activity. In addition to this, it has potential antibacterial activities against methicillin-resistant Staphylococcus aureus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloranilic acid is used in spectrophotometry. It reacts with metal ions to form stable complexes. It is also used as a reactant in the preparation of dimethylbipyridyl complexes and (nonylbenzimidazolylmethyl)benzene. Further, it is used in charge-transfer reactions with metoprolol tartrate. It is also employed in the synthesis of osmium metalla-rectangles with anticancer activity. In addition to this, it has potential antibacterial activities against methicillin-resistant Staphylococcus aureus.

Solubility
Soluble in hot water and methanol. Insoluble in organic solvents.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

87-88-7
208.978
IPPWILKGXFOXHO-UHFFFAOYSA-N
66604
C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O
C6H2Cl2O4
MFCD00001596
chloranilic acid, 2,5-dichloro-3,6-dihydroxy-p-benzoquinone, p-chloranilic acid, 2,5-cyclohexadiene-1,4-dione, 2,5-dichloro-3,6-dihydroxy, unii-yj8l3bb7y4, 2,5-dichloro-3,6-dihydroxybenzoquinone, 2,5-dihydroxy-3,6-dichlorobenzoquinone, yj8l3bb7y4, p-quinone, 2,5-dichloro-3,6-dihydroxy, p-benzoquinone, 2,5-dichloro-3,6-dihydroxy
2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione

Specifications

87-88-7
C6H2Cl2O4
1875040
chloranilic acid, 2,5-dichloro-3,6-dihydroxy-p-benzoquinone, p-chloranilic acid, 2,5-cyclohexadiene-1,4-dione, 2,5-dichloro-3,6-dihydroxy, unii-yj8l3bb7y4, 2,5-dichloro-3,6-dihydroxybenzoquinone, 2,5-dihydroxy-3,6-dichlorobenzoquinone, yj8l3bb7y4, p-quinone, 2,5-dichloro-3,6-dihydroxy, p-benzoquinone, 2,5-dichloro-3,6-dihydroxy
IPPWILKGXFOXHO-UHFFFAOYSA-N
2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione
66604
≥98%
>300°C (decomposition)
MFCD00001596
14,2079
Soluble in hot water and methanol. Insoluble in organic solvents.
C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O
208.978
208.99
Chloranilic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 201-780-7

missing translation for 'rtecsNumber' : DK4005000

missing translation for 'tsca' : Yes

Product Identifier
  • Chloranilic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Chloranilic acid

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.