missing translation for 'onlineSavingsMsg'
Learn More

Chloranilic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11458733
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chloranilic acid is used in spectrophotometry. It reacts with metal ions to form stable complexes. It is also used as a reactant in the preparation of dimethylbipyridyl complexes and (nonylbenzimidazolylmethyl)benzene. Further, it is used in charge-transfer reactions with metoprolol tartrate. It is also employed in the synthesis of osmium metalla-rectangles with anticancer activity. In addition to this, it has potential antibacterial activities against methicillin-resistant Staphylococcus aureus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloranilic acid is used in spectrophotometry. It reacts with metal ions to form stable complexes. It is also used as a reactant in the preparation of dimethylbipyridyl complexes and (nonylbenzimidazolylmethyl)benzene. Further, it is used in charge-transfer reactions with metoprolol tartrate. It is also employed in the synthesis of osmium metalla-rectangles with anticancer activity. In addition to this, it has potential antibacterial activities against methicillin-resistant Staphylococcus aureus.

Solubility
Soluble in hot water and methanol. Insoluble in organic solvents.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-88-7
Molecular Formula C6H2Cl2O4
Molecular Weight (g/mol) 208.978
MDL Number MFCD00001596
InChI Key IPPWILKGXFOXHO-UHFFFAOYSA-N
Synonym chloranilic acid, 2,5-dichloro-3,6-dihydroxy-p-benzoquinone, p-chloranilic acid, 2,5-cyclohexadiene-1,4-dione, 2,5-dichloro-3,6-dihydroxy, unii-yj8l3bb7y4, 2,5-dichloro-3,6-dihydroxybenzoquinone, 2,5-dihydroxy-3,6-dichlorobenzoquinone, yj8l3bb7y4, p-quinone, 2,5-dichloro-3,6-dihydroxy, p-benzoquinone, 2,5-dichloro-3,6-dihydroxy
PubChem CID 66604
IUPAC Name 2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione
SMILES C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O

Specifications

Melting Point >300°C (decomposition)
Quantity 100 g
Beilstein 1875040
Merck Index 14,2079
Solubility Information Soluble in hot water and methanol. Insoluble in organic solvents.
Formula Weight 208.99
Percent Purity ≥98%
Chemical Name or Material Chloranilic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'