missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Acetate, Extra Pure, SLR, Dried, Fisher Chemical™
97.95 € - 121.00 €
Chemical Identifiers
CAS | 62-54-4 |
---|---|
Molecular Formula | C4H6CaO4 |
Molecular Weight (g/mol) | 158.17 |
MDL Number | 12448 |
InChI Key | VSGNNIFQASZAOI-UHFFFAOYSA-L |
IUPAC Name | calcium diacetate |
SMILES | [Ca++].CC([O-])=O.CC([O-])=O |
Description
Chemical Identifiers
62-54-4 | |
158.17 | |
VSGNNIFQASZAOI-UHFFFAOYSA-L | |
[Ca++].CC([O-])=O.CC([O-])=O |
C4H6CaO4 | |
12448 | |
calcium diacetate |
Specifications
62-54-4 | |
7.6 | |
C4H6CaO4 | |
VSGNNIFQASZAOI-UHFFFAOYSA-L | |
calcium diacetate | |
158.17g/mol |
White | |
Solid | |
12448 | |
[Ca++].CC([O-])=O.CC([O-])=O | |
158.17 | |
Plastic powder jar |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title