missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Acetate, Extra Pure, SLR, Dried, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
Specifications
Specifications
Color | White |
pH | 7.6 |
Physical Form | Solid |
Quantity | 250 g |
Formula Weight | 158.17g/mol |
Packaging | Plastic powder jar |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction