Learn More
Cadmium selenide, 99.999% (metals basis), Thermo Scientific Chemicals
31.76 € - 538.68 €
Chemical Identifiers
CAS | 1306-24-7 |
---|---|
Molecular Formula | CdSe |
Molecular Weight (g/mol) | 191.39 |
MDL Number | MFCD00010917 |
InChI Key | AQCDIIAORKRFCD-UHFFFAOYSA-N |
Synonym | cadmium selenide, cadmium ii selenide, cadmium 2+ selenide, cadmium selenide nanotubes, cadmium selenide trace metals basis 1g, cadmium selenide candot quantum dot core , 50umol/l in hexane, lumidot tm cdse, 480, core-type quantum dots, 5 mg/ml in toluene, lumidot tm cdse, 520, core-type quantum dots, 5 mg/ml in toluene |
PubChem CID | 14784 |
ChEBI | CHEBI:50834 |
IUPAC Name | selanylidenecadmium |
SMILES | [Se]=[Cd] |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
15472267
|
Thermo Scientific Alfa Aesar
045456.04 |
2 g | N/A | N/A | N/A | |||||
15482267
|
Thermo Scientific Alfa Aesar
045456.09 |
10 g |
122.61 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15492267
|
Thermo Scientific Alfa Aesar
045456.18 |
50 g |
538.68 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Cadmium selenide is applied in opto-electronic devices, laser diodes, biomedical imaging, nanosensing, high-efficiency solar cells and thin-film transistors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1306-24-7 | |
191.39 | |
AQCDIIAORKRFCD-UHFFFAOYSA-N | |
14784 | |
selanylidenecadmium |
CdSe | |
MFCD00010917 | |
cadmium selenide, cadmium ii selenide, cadmium 2+ selenide, cadmium selenide nanotubes, cadmium selenide trace metals basis 1g, cadmium selenide candot quantum dot core , 50umol/l in hexane, lumidot tm cdse, 480, core-type quantum dots, 5 mg/ml in toluene, lumidot tm cdse, 520, core-type quantum dots, 5 mg/ml in toluene | |
CHEBI:50834 | |
[Se]=[Cd] |
Specifications
>1350°C | |
Black to Brown-Red | |
CdSe | |
UN3077 | |
cadmium selenide, cadmium ii selenide, cadmium 2+ selenide, cadmium selenide nanotubes, cadmium selenide trace metals basis 1g, cadmium selenide candot quantum dot core , 50umol/l in hexane, lumidot tm cdse, 480, core-type quantum dots, 5 mg/ml in toluene, lumidot tm cdse, 520, core-type quantum dots, 5 mg/ml in toluene | |
[Se]=[Cd] | |
191.39 | |
CHEBI:50834 | |
99.999% | |
5.81g/mL | |
Cadmium selenide |
1306-24-7 | |
Pieces | |
MFCD00010917 | |
14,1626 | |
AQCDIIAORKRFCD-UHFFFAOYSA-N | |
selanylidenecadmium | |
14784 | |
191.36 | |
Odorless | |
(metals basis) |
Safety and Handling
GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.