missing translation for 'onlineSavingsMsg'
Learn More

Cadmium selenide, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 15472267
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cadmium selenide is applied in opto-electronic devices, laser diodes, biomedical imaging, nanosensing, high-efficiency solar cells and thin-film transistors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-24-7
Molecular Formula CdSe
Molecular Weight (g/mol) 191.39
MDL Number MFCD00010917
InChI Key AQCDIIAORKRFCD-UHFFFAOYSA-N
Synonym cadmium selenide, cadmium ii selenide, cadmium 2+ selenide, cadmium selenide nanotubes, cadmium selenide trace metals basis 1g, cadmium selenide candot quantum dot core , 50umol/l in hexane, lumidot tm cdse, 480, core-type quantum dots, 5 mg/ml in toluene, lumidot tm cdse, 520, core-type quantum dots, 5 mg/ml in toluene
PubChem CID 14784
ChEBI CHEBI:50834
IUPAC Name selanylidenecadmium
SMILES [Se]=[Cd]

Specifications

Melting Point >1350°C
Color Black to Brown-Red
Physical Form Pieces
Quantity 2 g
UN Number UN3077
Merck Index 14,1626
Formula Weight 191.36
Percent Purity 99.999%
Odor Odorless
Density 5.81g/mL
Assay (metals basis)
Chemical Name or Material Cadmium selenide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.