Learn More
Cadmium hydroxide, 75%(as Cd), pure, Thermo Scientific™
65.22 € - 215.00 €
Chemical Identifiers
CAS | 21041-95-2 |
---|---|
Molecular Formula | CdH2O2 |
Molecular Weight (g/mol) | 146.43 |
MDL Number | MFCD00015995 |
InChI Key | PLLZRTNVEXYBNA-UHFFFAOYSA-L |
IUPAC Name | cadmium(2+) dihydroxide |
SMILES | [OH-].[OH-].[Cd++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15259896
|
Thermo Scientific Acros
453882500 |
250 g |
65.22 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15249896
|
Thermo Scientific Acros
453880010 |
1 kg | N/A | ||||||
Description
Chemical Identifiers
21041-95-2 | |
146.43 | |
PLLZRTNVEXYBNA-UHFFFAOYSA-L | |
[OH-].[OH-].[Cd++] |
CdH2O2 | |
MFCD00015995 | |
cadmium(2+) dihydroxide |
Specifications
232.0°C | |
75%(as Cd) | |
MFCD00015995 | |
Solubility in water: insoluble. Other solubilities: slightly sol in naoh sol, sol in dil acids, in nh4, or nh4cl sol | |
[OH-].[OH-].[Cd++] | |
146.43 | |
75% (as Cd) | |
Cadmium hydroxide |
21041-95-2 | |
CdH2O2 | |
15,1625 | |
PLLZRTNVEXYBNA-UHFFFAOYSA-L | |
cadmium(2+) dihydroxide | |
146.41 | |
Pure |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Suspected of causing genetic defects.
May cause cancer.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
Causes damage to organs through prolonged or repeated e
GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Use personal protective equipment as required.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 244-168-5
- Cadmium hydroxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
Hazard Statement
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H350-May cause cancer.
- H341-Suspected of causing genetic defects.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H330-Fatal if inhaled.
Precautionary Statement
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.