missing translation for 'onlineSavingsMsg'
Learn More

Cadmium hydroxide, 75%(as Cd), pure, Thermo Scientific™

Product Code. 15249896
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 21041-95-2
Molecular Formula CdH2O2
Molecular Weight (g/mol) 146.43
MDL Number MFCD00015995
InChI Key PLLZRTNVEXYBNA-UHFFFAOYSA-L
IUPAC Name cadmium(2+) dihydroxide
SMILES [OH-].[OH-].[Cd++]

Specifications

Melting Point 232.0°C
Quantity 1 kg
Assay Percent Range 75%(as Cd)
Merck Index 15,1625
Solubility Information Solubility in water: insoluble. Other solubilities: slightly sol in naoh sol, sol in dil acids, in nh4, or nh4cl sol
Formula Weight 146.41
Percent Purity 75% (as Cd)
Grade Pure
Chemical Name or Material Cadmium hydroxide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cadmium hydroxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350-May cause cancer.
  • H341-Suspected of causing genetic defects.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'