Learn More
Cadmium bromide tetrahydrate, 98%, pure, Thermo Scientific Chemicals
32.90 € - 270.00 €
Chemical Identifiers
CAS | 13464-92-1 |
---|---|
Molecular Formula | Br2Cd·4H2O |
Molecular Weight (g/mol) | 344.28 |
InChI Key | PANYFCXNZSEXSU-UHFFFAOYSA-L |
Synonym | cadmium dibromide, tetrahydrate, acmc-20alji, ksc497c4n, cadmium ii ; bromide tetrahydrate, cadmium 2+ dibromide tetrahydrate, cadmium 2+ ion tetrahydrate dibromide, cadmium bromide, tetrahydrate, reagent |
PubChem CID | 3084147 |
IUPAC Name | cadmium(2+);dibromide;tetrahydrate |
SMILES | O.O.O.O.[Br-].[Br-].[Cd+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15219896
|
Thermo Scientific Acros
453870250 |
25 g |
32.90 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15229896
|
Thermo Scientific Acros
453871000 |
100 g |
97.40 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15239896
|
Thermo Scientific Acros
453875000 |
500 g |
270.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13464-92-1 | |
344.28 | |
cadmium dibromide, tetrahydrate, acmc-20alji, ksc497c4n, cadmium ii ; bromide tetrahydrate, cadmium 2+ dibromide tetrahydrate, cadmium 2+ ion tetrahydrate dibromide, cadmium bromide, tetrahydrate, reagent | |
cadmium(2+);dibromide;tetrahydrate |
Br2Cd·4H2O | |
PANYFCXNZSEXSU-UHFFFAOYSA-L | |
3084147 | |
O.O.O.O.[Br-].[Br-].[Cd+2] |
Specifications
13464-92-1 | |
Br2Cd·4H2O | |
Solubility in water: soluble. | |
O.O.O.O.[Br-].[Br-].[Cd+2] | |
344.28 | |
344.28 | |
Pure |
97.5% min | |
cadmium dibromide, tetrahydrate, acmc-20alji, ksc497c4n, cadmium ii ; bromide tetrahydrate, cadmium 2+ dibromide tetrahydrate, cadmium 2+ ion tetrahydrate dibromide, cadmium bromide, tetrahydrate, reagent | |
PANYFCXNZSEXSU-UHFFFAOYSA-L | |
cadmium(2+);dibromide;tetrahydrate | |
3084147 | |
98% | |
Cadmium bromide tetrahydrate |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
GHS Signal Word: Warning
- Cadmium bromide, tetrahydrate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only