missing translation for 'onlineSavingsMsg'
Learn More

Cadmium bromide tetrahydrate, 98%, pure, Thermo Scientific Chemicals

Product Code. 15229896
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13464-92-1
Molecular Formula Br2Cd·4H2O
Molecular Weight (g/mol) 344.28
InChI Key PANYFCXNZSEXSU-UHFFFAOYSA-L
Synonym cadmium dibromide, tetrahydrate, acmc-20alji, ksc497c4n, cadmium ii ; bromide tetrahydrate, cadmium 2+ dibromide tetrahydrate, cadmium 2+ ion tetrahydrate dibromide, cadmium bromide, tetrahydrate, reagent
PubChem CID 3084147
IUPAC Name cadmium(2+);dibromide;tetrahydrate
SMILES O.O.O.O.[Br-].[Br-].[Cd+2]

Specifications

Quantity 100 g
Assay Percent Range 97.5% min
Solubility Information Solubility in water: soluble.
Formula Weight 344.28
Percent Purity 98%
Grade Pure
Chemical Name or Material Cadmium bromide tetrahydrate
compliance-icons compliance-icons
Product Identifier
  • Cadmium bromide, tetrahydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'