Learn More
Cadmium acetate, 98%, pure, anhydrous, Thermo Scientific Chemicals
116.00 € - 293.00 €
Chemical Identifiers
CAS | 543-90-8 |
---|---|
Molecular Formula | C4H6CdO4 |
Molecular Weight (g/mol) | 230.5 |
InChI Key | LHQLJMJLROMYRN-UHFFFAOYSA-L |
Synonym | cadmium acetate, cadmium diacetate, acetic acid, cadmium salt, cadmium ethanoate, bis acetoxy cadmium, cadmium di acetate, cadmium ii acetate, unii-95kc50z1l0, ccris 7783, hsdb 280 |
PubChem CID | 10986 |
IUPAC Name | cadmium(2+);diacetate |
SMILES | CC(=O)[O-].CC(=O)[O-].[Cd+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15229936
|
Thermo Scientific Acros
454002500 |
250 g |
116.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15219936
|
Thermo Scientific Acros
454000010 |
1 kg | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
543-90-8 | |
230.5 | |
cadmium acetate, cadmium diacetate, acetic acid, cadmium salt, cadmium ethanoate, bis acetoxy cadmium, cadmium di acetate, cadmium ii acetate, unii-95kc50z1l0, ccris 7783, hsdb 280 | |
cadmium(2+);diacetate |
C4H6CdO4 | |
LHQLJMJLROMYRN-UHFFFAOYSA-L | |
10986 | |
CC(=O)[O-].CC(=O)[O-].[Cd+2] |
Specifications
255.0°C to 273.0°C | |
97.5% min. (Complexometry) | |
15,1619 | |
LHQLJMJLROMYRN-UHFFFAOYSA-L | |
cadmium(2+);diacetate | |
10986 | |
98% | |
0.5% max. |
543-90-8 | |
C4H6CdO4 | |
cadmium acetate, cadmium diacetate, acetic acid, cadmium salt, cadmium ethanoate, bis acetoxy cadmium, cadmium di acetate, cadmium ii acetate, unii-95kc50z1l0, ccris 7783, hsdb 280 | |
CC(=O)[O-].CC(=O)[O-].[Cd+2] | |
230.5 | |
230.5 | |
Pure | |
Cadmium acetate |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause cancer.
GHS P Statement
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortab
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 208-853-2
- Cadmium acetate anhydrous
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1A
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H350-May cause cancer.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only