missing translation for 'onlineSavingsMsg'
Learn More

Cadmium acetate, 98%, pure, anhydrous, Thermo Scientific Chemicals

Product Code. 15219936
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-90-8
Molecular Formula C4H6CdO4
Molecular Weight (g/mol) 230.5
InChI Key LHQLJMJLROMYRN-UHFFFAOYSA-L
Synonym cadmium acetate, cadmium diacetate, acetic acid, cadmium salt, cadmium ethanoate, bis acetoxy cadmium, cadmium di acetate, cadmium ii acetate, unii-95kc50z1l0, ccris 7783, hsdb 280
PubChem CID 10986
IUPAC Name cadmium(2+);diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Cd+2]

Specifications

Melting Point 255.0°C to 273.0°C
Quantity 1 kg
Assay Percent Range 97.5% min. (Complexometry)
Merck Index 15,1619
Formula Weight 230.5
Percent Purity 98%
Grade Pure
Loss on Drying 0.5% max.
Chemical Name or Material Cadmium acetate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cadmium acetate anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350-May cause cancer.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.