missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C10orf68 Rabbit anti-Human, Non-human primate, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | C10orf68 |
---|---|
Concentration | 0.36 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883495
|
Proteintech
24975-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16873495
|
Proteintech
24975-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May play a role in tumorigenesis.Specifications
C10orf68 | |
Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Non-human Primate, Human | |
Q96M83 | |
79741 | |
C10orf68 Fusion Protein Ag21682 | |
Primary | |
-20°C | |
CCDC7 |
0.36 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C10orf68 | |
CCDC7 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title