missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C10orf68 Rabbit anti-Human, Non-human primate, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24975-1-AP-150UL
This item is not returnable.
View return policy
Description
May play a role in tumorigenesis.Specifications
C10orf68 | |
Polyclonal | |
Unconjugated | |
CCDC7 | |
C10orf68 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79741 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96M83 | |
CCDC7 | |
C10orf68 Fusion Protein Ag21682 | |
150 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction