missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution, 12.0, di-sodium hydrogen phosphate / sodium hydroxide solution, Honeywell Fluka™
di-Sodium hydrogen phosphate / sodium hydroxide solution
33.44 € - 351.97 €
Chemical Identifiers
CAS | 1336-21-6 |
---|---|
Molecular Formula | H5NO |
Molecular Weight (g/mol) | 35.05 |
MDL Number | MFCD00066650 |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
PubChem CID | 14923 |
ChEBI | CHEBI:18219 |
IUPAC Name | azanium;hydroxide |
SMILES | N.O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15694050
|
Honeywell-Fluka
33651-100ML |
100 mL |
33.44 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15614060
|
Honeywell Fluka
33651-6X100ML |
6 x 100 mL |
174.37 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15604060
|
Honeywell-Fluka
33651-1L |
1 L |
67.60 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15624060
|
Honeywell Fluka
33651-6X1L |
6 x 1 L |
351.97 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
1336-21-6 | |
35.05 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
CHEBI:18219 | |
N.O |
H5NO | |
MFCD00066650 | |
14923 | |
azanium;hydroxide |
Specifications
1336-21-6 | |
12.0 | |
MFCD00066650 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
azanium;hydroxide | |
14923 | |
Buffer solution |
1.00g/mL at 20°C | |
H5NO | |
NONH for all modes of transport | |
N.O | |
35.05 | |
CHEBI:18219 |