missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution, 12.0, di-sodium hydrogen phosphate / sodium hydroxide solution, Honeywell Fluka™
Click to view available options
Quantity:
100 mL
1 L
6 x 100 mL
6 x 1 L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Density | 1.00g/mL at 20°C |
Quantity | 6 x 100 mL |
UN Number | NONH for all modes of transport |
Chemical Name or Material | Buffer solution, di-sodium hydrogen phosphate / sodium hydroxide solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'