missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Purple Indicator, Reag. Ph. Eur., Honeywell Fluka™

Indicator, Reag. Ph. Eur.

51.62 € - 1479.46 €

Chemical Identifiers

CAS 115-40-2
Molecular Formula C21H16Br2O5S
Molecular Weight (g/mol) 540.22
MDL Number MFCD00011681
InChI Key ABIUHPWEYMSGSR-UHFFFAOYSA-N
PubChem CID 8273
ChEBI CHEBI:86154
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-6-methylphenol
SMILES CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15693820
View Documents Promotion Details
Honeywell-Fluka
32642-5G
5 g Glass Bottle
51.62 €
5g
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
 
15683820
View Documents Promotion Details
Honeywell-Fluka
32642-25G
25 g Glass Bottle
157.57 €
25g
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
 
15673820
View Documents Promotion Details
Honeywell-Fluka
32642-1KG
1 kg Glass Bottle
1479.46 €
1kg
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

115-40-2
540.22
ABIUHPWEYMSGSR-UHFFFAOYSA-N
CHEBI:86154
CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1
C21H16Br2O5S
MFCD00011681
8273
2-bromo-4-[3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-6-methylphenol

Specifications

240°C
C21H16Br2O5S
NONH for all modes of transport
ABIUHPWEYMSGSR-UHFFFAOYSA-N
2-bromo-4-[3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-6-methylphenol
8273
540.22g/mol
5.2 to 6.8 (yellow to purple)
115-40-2
MFCD00011681
359618
CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1
540.22
CHEBI:86154
Complies for change range
Bromocresol Purple, Reag. Ph. Eur.
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 204-087-8

SDS
Documents

Documents

Product Certifications