missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Purple Indicator, Reag. Ph. Eur., Honeywell Fluka™

Product Code. 15683820
Click to view available options
Quantity:
5 g
25 g
1 kg
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Indicator, Reag. Ph. Eur.

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-40-2
Molecular Formula C21H16Br2O5S
Molecular Weight (g/mol) 540.22
MDL Number MFCD00011681
InChI Key ABIUHPWEYMSGSR-UHFFFAOYSA-N
PubChem CID 8273
ChEBI CHEBI:86154
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-6-methylphenol
SMILES CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1

Specifications

Melting Point 240°C
UN Number NONH for all modes of transport
Beilstein 359618
Formula Weight 540.22g/mol
Packaging Glass Bottle
Suitability Complies for change range
pH 5.2 to 6.8 (yellow to purple)
Quantity 25 g
Chemical Name or Material Bromocresol Purple, Reag. Ph. Eur.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'