Learn More
Retsch™ Bottom Sieves SK 300 with Trapezoid Holes
Retsch™ Bottom Sieves SK 300 with Trapezoid Holes are constructed of stainless steel for durability and suitable for use with Cross Beater Mill SK 300.
294.00 €
Specifications
Description Trapezoid holes
Type Sieve
Material Stainless Steel
For Use With (Equipment) Cross Beater Mill SK 300
View More Specs
Products 1
Product Code Brand Pore Size Price Quantity & Availability  
Product Code Brand Pore Size Price Quantity & Availability  
15606455
View Documents Promotion Details
Retsch™
02.407.0100
250 μm
294.00 €
Each
Estimated Shipment: 11-02-2025
to see stock.
Add to basket
Description
Description
  • Stainless steel
  • Trapezoid holes
  • For use with cross beater
Specifications
Specifications
Trapezoid holes
Sieve
Stainless Steel
Cross Beater Mill SK 300