Learn More
Retsch™ Bottom Sieves SK 300 with Trapezoid Holes
Retsch™ Bottom Sieves SK 300 with Trapezoid Holes are constructed of stainless steel for durability and suitable for use with Cross Beater Mill SK 300.
Brand:  Retsch™ 02.407.0100
 View more versions of this product
Product Code. 15606455
  • 294.00 € / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stainless steel
  • Trapezoid holes
  • For use with cross beater
Specifications
Specifications
Trapezoid holes
Sieve
Cross Beater Mill SK 300
250 μm
Stainless Steel