Learn More
Karl Hecht™ Assistent™ Reduction and Expansion Adapters
For the quick and secure connection of glass joints in the laboratory. Karl Hecht™ Assistent™ Reduction and Expansion Adapters are manufactured from durable DURAN™ borosilicate 3.3 glass.
18.00 € - 48.00 €
Specifications
Material | Borosilicate Glass |
---|---|
Product Type | Duran sleeve |
Quantity | 1 Each |
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | |||||
10564042
|
Karl Hecht™
42387010 |
Socket size: 29/32 , Cone size: 14/23 |
23.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10155241
|
Karl Hecht™
42387013 |
Socket size: 29/32, Cone size: 24/29 |
29.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10297571
|
Karl Hecht™
42387014 |
Socket size: 45/40, Cone size: 29/32 |
48.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10629631
|
Karl Hecht™
42387012 |
Socket size: 29/32 , Cone size: 19/26 |
27.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10237291
|
Karl Hecht™
42387004 |
Socket size: 19/26, Cone size: 24/29 |
18.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10788102
|
Karl Hecht™
42387005 |
Socket size: 19/26, Cone size: 29/32 |
19.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10583462
|
Karl Hecht™
42387006 |
Socket size: 29/32, Cone size: 45/40 |
35.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10309451
|
Karl Hecht™
42387008 |
Socket size: 19/26, Cone size: 14/23 |
20.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Solid rim
- Acc. to DIN 12257
- Reinforced ground joint
Specifications
Borosilicate Glass | |
Duran sleeve | |
1 Each |