Learn More
Karl Hecht™ Assistent™ Reduction and Expansion Adapters
For the quick and secure connection of glass joints in the laboratory. Karl Hecht™ Assistent™ Reduction and Expansion Adapters are manufactured from durable DURAN™ borosilicate 3.3 glass.
Brand:  Karl Hecht™ 42387006
 View more versions of this product
Product Code. 10583462
  • 35.00 € / Each
Explore more special offers
Description
Description
  • Solid rim
  • Acc. to DIN 12257
  • Reinforced ground joint
Specifications
Specifications
Borosilicate Glass
Duran sleeve
1 Each
Socket size: 29/32, Cone size: 45/40