missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™

45.75 € - 154.00 €

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
MDL Number 4599
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10203470
View Documents
Fisher Chemical
B/2550/08
500 mL Amber glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10602261
View Documents
Fisher Chemical
B/2550/PB17
2.5 L Plastic Coated Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

100-51-6
C7H8O
4599

Specifications

100-51-6
Yellow
C7H8O
3.72
0.13mbar at 20°C
-15°C
205°C
4599
108.14g/mol
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Benzyl alcohol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications