missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Alcohol, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Amber glass bottle
Plastic Coated Glass bottle
Specifications
Specifications
Melting Point | -15°C |
Color | Yellow |
Boiling Point | 205°C |
Packaging | Plastic Coated Glass bottle |
Quantity | 2.5 L |
Vapor Density | 3.72 |
Formula Weight | 108.14g/mol |
Vapor Pressure | 0.13mbar at 20°C |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Benzyl alcohol
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'