Learn More
Cole-Parmer™ PVDF Union T-Connector Compression Fittings
Compression fittings work well with rigid tubing for more permanent installation in high-pressure applications.
Brand: Cole-Parmer™ 06382-50
This item is not returnable.
View return policy
Description
- No leaks, flow restrictions or costly downtime
- Fluid paths in the laboratory assembled without the use of tools
- Provided with nut that compresses rigid tubing against fitting body
- Sterilization by autoclave or ethylene oxide (ETO)
- Offering excellent chemical resistance
Specifications
Compression Fitting | |
PVDF | |
1 Each | |
7/32 in. orifice; 1-15/32 x 1-1/32 in. | |
26 mm |
Autoclavable | |
1/4 in. tubing O.D. | |
Excellent chemical resistance | |
Autoclave or Ethylene Oxide |