missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Polysulfide Solution, Certified AR for Analysis, Fisher Chemical™

29.00 € - 130.00 €

Chemical Identifiers

MDL Number 242670
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10650311
View Documents Promotion Details
Fisher Chemical
A/6560/PB08
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10539760
View Documents Promotion Details
Fisher Chemical
A/6560/PB17
2.5 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Chemical Identifiers

242670

Specifications

Yellow
Liquid
242670
Plastic Coated Glass bottle
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ammonium polysulfide solution

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.
  • EUH031-Contact with acids liberates toxic gas.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.

Supplemental information
  • MIXTURE LIST-Contains : Ammonium polysulfide

SDS
Documents

Documents

Product Certifications