missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Polysulfide Solution, Certified AR for Analysis, Fisher Chemical™

Product Code. 10539760
Click to view available options
Quantity:
500 mL
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

MDL Number 242670

Specifications

Color Yellow
Physical Form Liquid
Quantity 2.5 L
Packaging Plastic Coated Glass bottle
compliance-icons compliance-icons
Product Identifier
  • AMMONIUM SULFIDE
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'