missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Phosphate Monobasic, ACS Reagent, Honeywell Fluka™
ACS Reagent,≥98%
34.79 € - 366.00 €
Chemical Identifiers
CAS | 7722-76-1 |
---|---|
Molecular Formula | H6NO4P |
Molecular Weight (g/mol) | 115.03 |
MDL Number | MFCD00003396 |
InChI Key | LFVGISIMTYGQHF-UHFFFAOYSA-N |
Synonym | ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate |
PubChem CID | 24402 |
ChEBI | CHEBI:62982 |
IUPAC Name | azanium;dihydrogen phosphate |
SMILES | N.OP(O)(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15612020
|
Honeywell-Fluka
216003-100G |
100 g | N/A | ||||||
15632020
|
Honeywell-Fluka
216003-500G |
500 g |
154.13 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15622020
|
Honeywell-Fluka
216003-2.5KG |
2.5 kg | N/A | ||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7722-76-1 | |
115.03 | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
24402 | |
azanium;dihydrogen phosphate |
H6NO4P | |
MFCD00003396 | |
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate | |
CHEBI:62982 | |
N.OP(O)(O)=O |
Specifications
7722-76-1 | |
NH4H2PO4 | |
NONH for all modes of transport | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
azanium;dihydrogen phosphate | |
24402 | |
115.03g/mol | |
ACS Reagent |
H6NO4P | |
MFCD00003396 | |
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate | |
N.OP(O)(O)=O | |
115.03 | |
CHEBI:62982 | |
≥98% | |
Ammonium Phosphate Monobasic |
Safety and Handling
missing translation for 'einecsNumber' : 231-764-5
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title