missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate Monobasic, ACS Reagent, Honeywell Fluka™

Product Code. 15612020
Click to view available options
Quantity:
100 g
500 g
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
PubChem CID 24402
ChEBI CHEBI:62982
IUPAC Name azanium;dihydrogen phosphate
SMILES N.OP(O)(O)=O

Specifications

Quantity 100 g
Linear Formula NH4H2PO4
UN Number NONH for all modes of transport
Formula Weight 115.03g/mol
Percent Purity ≥98%
Grade ACS Reagent
Chemical Name or Material Ammonium Phosphate Monobasic
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'