missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amberlyst™ 15, (dry) ion-exchange resin, Thermo Scientific Chemicals
27.70 € - 373.00 €
Chemical Identifiers
CAS | 9037-24-5 |
---|---|
Molecular Formula | MFCD00145841 |
Molecular Weight (g/mol) | 0.00 |
MDL Number | MFCD00145841 |
InChI Key | YZUPZGFPHUVJKC-UHFFFAOYSA-N |
PubChem CID | 80972 |
SMILES | * |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10286540
|
Thermo Scientific Acros
202140250 |
25 g | Plastic bottle |
27.70 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10490091
|
Thermo Scientific Acros
202145000 |
500 g | Plastic bottle |
114.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10711424
|
Thermo Scientific Acros
202140025 |
2.5 kg | Plastic bottle |
373.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resinChemical Identifiers
9037-24-5 | |
0.00 | |
YZUPZGFPHUVJKC-UHFFFAOYSA-N | |
* |
MFCD00145841 | |
MFCD00145841 | |
80972 |
Specifications
Beads | |
Particle size analysis:% <0.300mm: <1.0% | |
MFCD00145841 | |
15, 377 | |
* | |
80972 |
9037-24-5 | |
MFCD00145841 | |
05,355; 06,302; 09,256 | |
YZUPZGFPHUVJKC-UHFFFAOYSA-N | |
0.00 | |
Amberlyst™ 15 |
Safety and Handling
Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Amberlyst 15 ion-exchange resin
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only