missing translation for 'onlineSavingsMsg'
Learn More

Amberlyst™ 15, (dry) ion-exchange resin, Thermo Scientific Chemicals

Product Code. 10711424
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9037-24-5
Molecular Formula MFCD00145841
Molecular Weight (g/mol) 0.00
MDL Number MFCD00145841
InChI Key YZUPZGFPHUVJKC-UHFFFAOYSA-N
PubChem CID 80972
SMILES *

Specifications

Physical Form Beads
Particle Size Particle size analysis:% <0.300mm: <1.0%
Packaging Plastic bottle
Fieser 05,355; 06,302; 09,256
Merck Index 15, 377
Quantity 2.5 kg
Chemical Name or Material Amberlyst™ 15

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'