missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Potassium Sulfate, Extra Pure, SLR, Fisher Chemical™

48.40 € - 450.00 €

Chemical Identifiers

CAS 7784-24-9
MDL Number 149143
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11443713
View Documents
Fisher Chemical
A/2400/53
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10765321
View Documents
Fisher Chemical
A/2400/60
1 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10588410
View Documents
Fisher Chemical
A/2400/63
3 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Aluminum Potassium Sulfate, Extra Pure, SLR

Specifications

Chemical Identifiers

7784-24-9
149143

Specifications

92°C
White
Solid
149143
Plastic powder jar
7784-24-9
3.0 to 3.5
AlKO8S2·12H2O
474.39g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Aluminium potassium sulfate dodecahydrate

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.